Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB11 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989786
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9786 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9786 Supplier Bioss Supplier No. BS14369R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJB11 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity bystimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus a glycine/phenylalanine (G/F)-rich region and a C-terminal cysteine-rich region.

Specifications

Antigen DNAJB11
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene DNAJB11
Gene Accession No. Q9UBS4
Gene Alias 1810031F23Rik; ABBP2; ABBP-2; AL024055; APOBEC1-binding protein 2; BOS_919; cb954; DJ9; Dj-9; DnaJ (Hsp40) homolog, subfamily B, member 11; DnaJ heat shock protein family (Hsp40) member B11; DnaJ homolog subfamily B member 11; DnaJ protein 9; DnaJ protein homolog 9; DNAJB11; dnajb11 protein; EDJ; Endoplasmic reticulum DNA J domain-containing protein 3; ER-associated DNAJ; ER-associated dnaJ protein 3; ER-associated Hsp40 co-chaperone; ERdj3; ERJ3; ERj3p; ER-resident protein ERdj3; HDJ9; hDj-9; HEDJ; Human DnaJ protein 9; Liver regeneration-related protein LRRGT00084; LRRGT00084; PRO1080; PSEC0121; PWP1-interacting protein 4; UNQ537; UNQ537/PRO1080
Gene Symbols DNAJB11
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNAJB11.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51726
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.