Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJB12 Polyclonal Antibody
DNAJB12 Polyclonal Antibody
Supplier: Thermo Scientific PA568842
Description
DNAJB12 Polyclonal Antibody for Western Blot

Specifications
DNAJB12 | |
Polyclonal | |
Unconjugated | |
DNAJB12 | |
DJ10; DKFZp586B2023; DnaJ (Hsp40) homolog, subfamily B, member 12; DnaJ heat shock protein family (Hsp40) member B12; DnaJ homolog 10; dnaJ homolog subfamily B member 12; DnaJ subfamily B member 12-like protein; Dnajb12; LOW QUALITY PROTEIN: dnaJ homolog subfamily B member 12; mDj10 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
56709 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9QYI4 | |
DNAJB12 | |
synthetic peptide corresponding to a region of mouse DNAJB12 (aa 51-100). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction