Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB12 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983257
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3257 Supplier Bioss Supplier No. BS14371R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJB12 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000).

Specifications

Antigen DNAJB12
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene DNAJB12
Gene Accession No. Q9NXW2
Gene Alias BOS_24213; DJ10; DKFZp586B2023; DnaJ (Hsp40) homolog, subfamily B, member 12; DnaJ heat shock protein family (Hsp40) member B12; DnaJ homolog 10; DnaJ homolog subfamily B member 12; DnaJ subfamily B member 12-like protein; Dnajb12; mDj10
Gene Symbols DNAJB12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNAJB12.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54788
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.