Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562617
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562617 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562617 Supplier Invitrogen™ Supplier No. PA562617
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVKPGWRQGT RITFEKEGDQ GPNIIPADII FIVKEKLHPR FRRENDNLFF VNPIPLGKAL TCCTVEVRTL DDRLLNIPIN DIIHPKYFKK VPGE Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

DNAJA13 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB13
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJB13
Gene Accession No. P59910
Gene Alias 1700014P03Rik; DnaJ (Hsp40) homolog, subfamily B, member 13; DnaJ (Hsp40) related, subfamily B, member 13; DnaJ heat shock protein family (Hsp40) member B13; dnaJ homolog subfamily B member 13; DNAJB13; DnaJ-like protein; radial spoke 16 homolog A; RSPH16A; spermatogenesis apoptosis-related protein; testis and spermatogenesis cell-related protein 6; Testis spermatocyte apoptosis-related gene 6 protein; testis spermatogenesis apoptosis related protein 1; testis spermatogenesis apoptosis-related gene 3 protein; Testis spermatogenesis apoptosis-related gene 6 protein; testis spermatogenesis apoptosis-related protein 1; testis spermatogenesis apoptosis-related protein 6; Tsarg; Tsarg1; TSARG3; TSARG5; TSARG6
Gene Symbols DNAJB13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJB13. Recombinant protein control fragment (Product #RP-97544).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374407
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.