Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJB14 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297528R
Description
DNAJB14 Polyclonal specifically detects DNAJB14 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
DNAJB14 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
DnaJ (Hsp40) homolog, subfamily B, member 14, dnaJ homolog subfamily B member 14, EGNR9427, FLJ14281, MGC22187, PRO34683 | |
Produced in rabbits immunized with E. coli-derived Human DNAJB14 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
79982 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction