Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117007
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117007 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117007 Supplier Invitrogen™ Supplier No. PA5117007
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: 293T, Jurkat, HeLa, MCF7, Mouse brain, Rat brain Immunogen sequence: MEGNRDEAEK CVEIAREALN AGNREKAQRF LQKAEKLYPL PSARALLEII MKNGSTAGNS PHCRKPSGSG DQSKPNCTKD STSGSGEGGK GYTKDQVDGV LSINKCKNYY EVLGVTKDAG.

Acts as a co-chaperone with HSPA8/Hsc70; required to promote protein folding and trafficking, prevent aggregation of client proteins, and promote unfolded proteins to endoplasmic reticulum-associated degradation (ERAD) pathway (PubMed:24732912). Acts by determining HSPA8/Hsc70's ATPase and polypeptide-binding activities (PubMed:24732912). Can also act independently of HSPA8/Hsc70: together with DNAJB12, acts as a chaperone that promotes maturation of potassium channels KCND2 and KCNH2 by stabilizing nascent channel subunits and assembling them into tetramers (PubMed:27916661). While stabilization of nascent channel proteins is dependent on HSPA8/Hsc70, the process of oligomerization of channel subunits is independent of HSPA8/Hsc70 (PubMed:27916661). When overexpressed, forms membranous structures together with DNAJB12 and HSPA8/Hsc70 within the nucleus; the role of these structures, named DJANGOs, is still unclear (PubMed:24732912). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB14
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.22 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dnajb14
Gene Accession No. Q149L6, Q8TBM8
Gene Alias 5730496F10Rik; DnaJ (Hsp40) homolog, subfamily B, member 14; DnaJ heat shock protein family (Hsp40) member B14; dnaJ homolog subfamily B member 14; Dnajb14; EGNR9427; PRO34683; UNQ9427/PRO34683
Gene Symbols Dnajb14
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human DNAJB14 (NP_0010268931).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499716, 70604, 79982
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.