Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJB2 Antibody (OTI4D12), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270951UV
Description
DNAJB2 Monoclonal specifically detects DNAJB2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
DNAJB2 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DNAJB2 | |
Full length recombinant protein of human DNAJB2 (NP_001034639) produced in HEK293T cells. | |
0.1 mL | |
3300.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
OTI4D12 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
DnaJ (Hsp40) homolog, subfamily B, member 2, HSJ-1, HSPF3neuronal DNAJ-like 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction