Learn More
Invitrogen™ DNAJB2 Polyclonal Antibody

Description
Recommended positive controls: A431. Predicted reactivity: Mouse (91%), Rat (83%), Rhesus Monkey (100%), Chimpanzee (99%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | DNAJB2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | DNAJB2 |
| Gene Accession No. | P25686 |
| Gene Alias | 2700059H22Rik; CMT2T; DnaJ (Hsp40) homolog, subfamily B, member 10; DnaJ (Hsp40) homolog, subfamily B, member 2; DnaJ heat shock protein family (Hsp40) member B2; dnaJ homolog subfamily B member 10; DnaJ homolog subfamily B member 2; dnaJ protein homolog 1; Dnajb10; DNAJB2; DSMA5; Heat shock 40 kDa protein 3; heat shock protein J1; heat shock protein, DNAJ-like 1; heat shock protein, neuronal DNAJ-like 1; HSJ1; HSJ-1; HSPF3; mDj8 |
| Gene Symbols | DNAJB2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.