Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAJB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540117
Description
Recommended positive controls: K562 cells. Predicted reactivity: Bovine (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
May operate as a co-chaperone of the male germ cell- and haploid stage-specific Hsp70 proteins. [UniProt].
Specifications
DNAJB3 | |
Polyclonal | |
Unconjugated | |
DNAJB3 | |
DnaJ (Hsp40) homolog, subfamily B, member 3; DnaJ heat shock protein family (Hsp40) member B3; dnaJ homolog subfamily B member 3; DnaJ Hsp40 homolog; DnaJ protein homolog 3; DNAJB3; HCG3; heat shock protein J3; heat shock protein, DNAJ-like 3; Hsj3; HSJ-3; Msj1; MSJ-1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
414061 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.75 mg/mL | |
0.1M Tris glycine with 20% glycerol, 0.1M glycine and 0.01% Thimerosal; pH 7 | |
Q8WWF6 | |
DNAJB3 | |
Recombinant protein encompassing a sequence within the center region of human DNAJB3. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction