Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB5 Monoclonal Antibody (OTI1B12)
GREENER_CHOICE

Catalog No. PIMA527373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527373 Supplier Invitrogen™ Supplier No. MA527373
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNAJB5 Monoclonal Antibody for Western Blot

DNAJB5 belongs to the evolutionarily conserved DNAJ/HSP40 protein family. For background information on the DNAJ family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB5
Applications Western Blot
Classification Monoclonal
Clone OTI1B12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNAJB5
Gene Accession No. O75953
Gene Alias 1110058L06Rik; AI462558; DnaJ (Hsp40) homolog, subfamily B, member 5; DnaJ heat shock protein family (Hsp40) member B5; dnaJ homolog subfamily B member 5; DNAJB5; heat shock cognate 40; heat shock protein cognate 40; Heat shock protein Hsp40-2; heat shock protein hsp40-3; Hsc40; Hsp40-3
Gene Symbols DNAJB5
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 1-261 of DNAJB5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25822
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.