Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB5 Monoclonal Antibody (OTI6F11), OriGene
SDP

Catalog No. 502543998
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3998 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3998 Supplier OriGene Supplier No. CF810909
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

DNAJB5 Monoclonal antibody specifically detects DNAJB5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen DNAJB5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6F11
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene AZIN1
Gene Accession No. O14977
Gene Alias 1110058L06Rik; AI462558; DnaJ (Hsp40) homolog, subfamily B, member 5; DnaJ heat shock protein family (Hsp40) member B5; dnaJ homolog subfamily B member 5; DNAJB5; heat shock cognate 40; heat shock protein cognate 40; Heat shock protein Hsp40-2; heat shock protein hsp40-3; Hsc40; Hsp40-3
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 260-448 of human AZIN1 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51582
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.