Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103407
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103407 Supplier Invitrogen™ Supplier No. PA5103407
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DNAJB5.

DNAJB5 belongs to the evolutionarily conserved DNAJ/HSP40 protein family. For background information on the DNAJ family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB5
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene DNAJB5
Gene Accession No. O75953, O89114
Gene Alias 1110058L06Rik; AI462558; DnaJ (Hsp40) homolog, subfamily B, member 5; DnaJ heat shock protein family (Hsp40) member B5; dnaJ homolog subfamily B member 5; DNAJB5; heat shock cognate 40; heat shock protein cognate 40; Heat shock protein Hsp40-2; heat shock protein hsp40-3; Hsc40; Hsp40-3
Gene Symbols DNAJB5
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAJB5(Accession O75953), corresponding to amino acid residues K209-T259.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25822, 56323
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.