Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584031
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584031 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584031 Supplier Invitrogen™ Supplier No. PA584031
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IDGRVIPLPC NDVIKPGTVK RLRGEGLPFP KVPTQRGDLI VEFKVRFPDR LTPQTRQILK QHLPCS Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

DNAJB5 belongs to the evolutionarily conserved DNAJ/HSP40 protein family. For background information on the DNAJ family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB5
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJB5
Gene Accession No. O75953
Gene Alias 1110058L06Rik; AI462558; DnaJ (Hsp40) homolog, subfamily B, member 5; DnaJ heat shock protein family (Hsp40) member B5; dnaJ homolog subfamily B member 5; DNAJB5; heat shock cognate 40; heat shock protein cognate 40; Heat shock protein Hsp40-2; heat shock protein hsp40-3; Hsc40; Hsp40-3
Gene Symbols DNAJB5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJB5. Recombinant protein control fragment (Product #RP-106314).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25822
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.