Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJB6 Mouse anti-Human, Mouse, Rat, Clone: OTI3H4, lyophilized, TrueMAB™
SDP

Catalog No. 501670233
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-0233 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-0233 Supplier OriGene Supplier No. CF810660
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

In prokaryotes, the heat shock response is characterized by the induction of several proteins including DnaJ, DnaK, GroEL and GroES. DnaK is a member of the HSP70 family, and like other members, exhibits ATPase activity which facilitates the release of bound proteins and disaggregation of protein complexes within the cell. Members of this protein group serve as molecular chaperones. Levels of DnaJ and DnaK are closely linked and, in E. coli, are constituents of a single operon. DnaJ regulates DnaK in several different systems. Relative to DnaK, the level of DnaJ synthesis is quite low. Studies have shown that DnaJ and DnaK, together with GrpE, cooperatively interact with firefly luciferase as it unfolds during thermal denaturation allowing renaturation of the protein upon return to normal temperatures. Also, DnaK together with DnaJ stimulates the binding of RepA initiator protein to the origin of P1 phage in vitro.

Specifications

Antigen DNAJB6
Applications Western Blot
Classification Monoclonal
Clone OTI3H4
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene DNAJB6
Gene Accession No. O54946, O75190, Q6AYU3
Gene Alias DJ4, DnaJ, HHDJ1, HSJ-2, HSJ2, LGMD1D, LGMD1E, MRJ, MSJ-1
Gene Symbols Dnajb6
Host Species Mouse
Immunogen Full length human recombit protein of human DNAJB6 produced in HEK293T.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10049, 23950, 362293
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.