Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAJB7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551405
Description
Immunogen sequence: VLGLQRYASP EDIKKAYHKV ALKWHPDKNP ENKEEAERKF KEVAEAYEVL SNDEKRDIYD KYGTEGLNGG GSHFDDECEY GFTFHKPDDV FKEIFHERDP FSFHFFEDSL EDLLNRPGSS YGNRNRDAGY FFSTASEYP Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 80%.
DNAJB7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 ).
Specifications
DNAJB7 | |
Polyclonal | |
Unconjugated | |
Dnajb7 | |
4933424H20Rik; DJ5; DnaJ (Hsp40) homolog, subfamily B, member 7; DnaJ heat shock protein family (Hsp40) member B7; dnaJ homolog subfamily B member 7; Dnajb7; dnaJ-like 5; HSC3; mDj5; novel DnaJ domain protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
150353 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z6W7 | |
Dnajb7 | |
Recombinant protein corresponding to Human DNAJB7. Recombinant protein control fragment (Product #RP-91387). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction