Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559621
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559621 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559621 Supplier Invitrogen™ Supplier No. PA559621
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TLSDANRRKE YDTLGHSAFT SGKGQRGSGS SFEQSFNFNF DDLFKDFGFF GQNQNTGSKK RFENHFQTRQ DG Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 86%.

This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB9
Applications Immunohistochemistry (Paraffin), Immunohistochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJB9
Gene Accession No. Q9UBS3
Gene Alias AA408011; AA673251; AA673481; AW556981; DKFZP564F1862; DNA j protein b9; DnaJ (Hsp40) homolog, subfamily B, member 9; DnaJ heat shock protein family (Hsp40) member B9; dnaJ homolog subfamily B member 9; dnaJ homolog, subfamily b, member 9; DNAJB9; Endoplasmic reticulum DNA J domain-containing protein 4; endoplasmic reticulum DnaJ homolog 4; ERdj4; ER-resident protein ERdj4; Mdg1; MDG-1; mDj7; microvascular endothelial differentiation gene 1; microvascular endothelial differentiation gene 1 protein; MST049; MSTP049; UNQ743/PRO1471
Gene Symbols DNAJB9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJB9. Recombinant protein control fragment (Product #RP-98421).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4189
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.