Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596702 Supplier Invitrogen™ Supplier No. PA596702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKSYYDILGV PKSASERQIK KAFHKLAMKY HPDKNKSPDA EAKFREIAEA YETLSDANRR KEYDTLGHSA FTSGKGQRGS GSSFEQSFNF NFDDLFKDFG FFGQNQNTGS KKRFENHFQT RQDGGSSRQR HHFQEFSFGG GLFDDMFEDM EKMFSFSGFD STNQHTVQTE NRFHGSSKHC RTVTQRRGNM VTTYTDCSGQ; Positive Samples: Mouse liver, Rat liver; Cellular Location: Endoplasmic reticulum lumen.

This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DNAJB9
Gene Accession No. P97554, Q9QYI6
Gene Alias AA408011; AA673251; AA673481; AW556981; DKFZP564F1862; DNA j protein b9; DnaJ (Hsp40) homolog, subfamily B, member 9; DnaJ heat shock protein family (Hsp40) member B9; dnaJ homolog subfamily B member 9; dnaJ homolog, subfamily b, member 9; DNAJB9; Endoplasmic reticulum DNA J domain-containing protein 4; endoplasmic reticulum DnaJ homolog 4; ERdj4; ER-resident protein ERdj4; Mdg1; MDG-1; mDj7; microvascular endothelial differentiation gene 1; microvascular endothelial differentiation gene 1 protein; MST049; MSTP049; UNQ743/PRO1471
Gene Symbols DNAJB9
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24908, 27362
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.