Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJB9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142776
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142776 Supplier Invitrogen™ Supplier No. PA5142776
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:16,000.

This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJB9
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene DNAJB9
Gene Accession No. Q9QYI6, Q9UBS3
Gene Alias AA408011; AA673251; AA673481; AW556981; DKFZP564F1862; DNA j protein b9; DnaJ (Hsp40) homolog, subfamily B, member 9; DnaJ heat shock protein family (Hsp40) member B9; dnaJ homolog subfamily B member 9; dnaJ homolog, subfamily b, member 9; DNAJB9; Endoplasmic reticulum DNA J domain-containing protein 4; endoplasmic reticulum DnaJ homolog 4; ERdj4; ER-resident protein ERdj4; Mdg1; MDG-1; mDj7; microvascular endothelial differentiation gene 1; microvascular endothelial differentiation gene 1 protein; MST049; MSTP049; UNQ743/PRO1471
Gene Symbols DNAJB9
Host Species Goat
Immunogen Peptide with sequence C-PDAEAKFREIAEAYE (aa 61-75).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27362, 4189
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.