Learn More
Invitrogen™ DNAJB9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528137
Description
Recommended positive controls: MDG1 tansfected 293T cell. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene is a member of the J protein family. J proteins function in many cellular processes by regulating the ATPase activity of 70 kDa heat shock proteins. This gene is a member of the type 2 subgroup of DnaJ proteins. The encoded protein is localized to the endoplasmic reticulum. This protein is induced by endoplasmic reticulum stress and plays a role in protecting stressed cells from apoptosis.
Specifications
| DNAJB9 | |
| Polyclonal | |
| Unconjugated | |
| DNAJB9 | |
| AA408011; AA673251; AA673481; AW556981; DKFZP564F1862; DNA j protein b9; DnaJ (Hsp40) homolog, subfamily B, member 9; DnaJ heat shock protein family (Hsp40) member B9; dnaJ homolog subfamily B member 9; dnaJ homolog, subfamily b, member 9; DNAJB9; Endoplasmic reticulum DNA J domain-containing protein 4; endoplasmic reticulum DnaJ homolog 4; ERdj4; ER-resident protein ERdj4; Mdg1; MDG-1; mDj7; microvascular endothelial differentiation gene 1; microvascular endothelial differentiation gene 1 protein; MST049; MSTP049; UNQ743/PRO1471 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4189 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.32 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q9UBS3 | |
| DNAJB9 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human DNAJB9. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.