Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJC1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP232127R
Description
DNAJC1 Polyclonal antibody specifically detects DNAJC1 in Human, Mouse samples. It is validated for Western BlotSpecifications
DNAJC1 | |
Polyclonal | |
Western Blot | |
DnaJ (Hsp40) homolog, subfamily C, member 1, dnaJ homolog subfamily C member 1, DnaJ protein homolog MTJ1, DNAJL1, DnaJ-like protein, ERdj1, HTJ1, MGC131954, MTJ1 | |
The immunogen this antibody was made to, maps to a region between residue 475 to 525 of human DnaJ (Hsp40) Homolog, Subfamily C, Member 1 using the numbering given in entry NP_071760.2 (GeneID 64215). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
64215 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction