Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553059
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553059 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553059 Supplier Invitrogen™ Supplier No. PA553059
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EDLSQLTRSM VKFPGGTPGR WEKIAHELGR SVTDVTTKAK QLKDSVTCSP GMVRLSELKS TVQNSRPIKT ATTLPDDMIT QREDAEGVAA EEEQEGDSGE QETGATDARP RRRKPARLLE ATAKPEPEEK SRAKRQKDFD IAEQNESS Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 75%.

The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC1
Gene Accession No. Q96KC8
Gene Alias 4733401K02Rik; AA960110; D230036H06Rik; DnaJ (Hsp40) homolog, subfamily C, member 1; DnaJ (Hsp40) homolog, subfamily C, member 1-like; DnaJ heat shock protein family (Hsp40) member C1; dnaJ homolog subfamily C member 1; DnaJ protein homolog MTJ1; Dnajc1; Dnajl1; DnaJ-like protein; DnaJ-like protein 1; ERdj1; ERj1p; HTJ1; MTJ1
Gene Symbols DNAJC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJC1. Recombinant protein control fragment (Product #RP-90265).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64215
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.