Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143983
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143983 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143983 Supplier Invitrogen™ Supplier No. PA5143983
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human HELA whole cell, human HEPG2 whole cell, human U87 whole cell, rat stomach tissue, rat RH35 whole cell, mouse stomach tissue, mouse HEPA1-6 whole cell. IHC: human breast cancer tissue, human placenta tissue, human prostate cancer tissue, human gastric cancer tissue. ICC/IF: A431 cell. Flow: HELA cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

This endoplasmic reticulum co-chaperone may play a role in protein folding and translocation across the endoplasmic reticulum membrane. DNAJC10 may act as a co-chaperone for HSPA5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC10
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene Dnajc10
Gene Accession No. Q498R3, Q8IXB1, Q9DC23
Gene Alias 1200006L06Rik; D2Ertd706e; DnaJ (Hsp40) homolog, subfamily C, member 10; DnaJ heat shock protein family (Hsp40) member C10; dnaJ homolog subfamily C member 10; DNAJC10; DnaJ-like protein subfamily C member 10-like protein; Endoplasmic reticulum DNA J domain-containing protein 5; endoplasmic reticulum DnaJ-PDI fusion protein 1; ERdj5; ER-resident protein ERdj5; J domain-containing PDI-like protein; j domain-containing protein disulfide isomerase-like protein; J-domain-containing protein disulfide isomerase-like protein; Jpdi; Macrothioredoxin; MTHr; PDIA19; protein disulfide isomerase family A, member 19; UNQ495/PRO1012; zgc:162218
Gene Symbols Dnajc10
Host Species Rabbit
Immunogen E.coli-derived human DNAJC10 recombinant protein (Position: D33-L793).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295690, 54431, 66861
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.