Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556645 Supplier Invitrogen™ Supplier No. PA556645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LNSLDAKEIY LEVIHNLPDF ELLSANTLED RLAHHRWLLF FHFGKNENSN DPELKKLKTL LKNDHIQVGR FDCSSAPDIC SNLYVFQPSL AVFKGQGTKE YEIHHGKKIL YDILAFAKES VNSHVTTLGP QNFPANDKEP WLVDF Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 92%.

This endoplasmic reticulum co-chaperone may play a role in protein folding and translocation across the endoplasmic reticulum membrane. DNAJC10 may act as a co-chaperone for HSPA5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dnajc10
Gene Accession No. Q8IXB1
Gene Alias 1200006L06Rik; D2Ertd706e; DnaJ (Hsp40) homolog, subfamily C, member 10; DnaJ heat shock protein family (Hsp40) member C10; dnaJ homolog subfamily C member 10; DNAJC10; DnaJ-like protein subfamily C member 10-like protein; Endoplasmic reticulum DNA J domain-containing protein 5; endoplasmic reticulum DnaJ-PDI fusion protein 1; ERdj5; ER-resident protein ERdj5; J domain-containing PDI-like protein; j domain-containing protein disulfide isomerase-like protein; J-domain-containing protein disulfide isomerase-like protein; Jpdi; Macrothioredoxin; MTHr; PDIA19; protein disulfide isomerase family A, member 19; UNQ495/PRO1012; zgc:162218
Gene Symbols Dnajc10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJC10. Recombinant protein control fragment (Product #RP-89989).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54431
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.