Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588377
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588377 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588377 Supplier Invitrogen™ Supplier No. PA588377
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: IDCQQYHSFC AQENVQRYPE IRFFPPKSNK AYHYHSYNGW NRDAYSLRIW GLGFLPQVST DLTPQTFSEK VLQGKNHWVI DFYAPWCGPC QNFAPEFELL ARMIKGKVKA GKVDCQAYAQ TCQKAGIRAY PTVKFYFYER AKRNFQEEQI NTRDAKAIAA LISEKLETLR NQGKRNKDEL; Positive Samples: U-87MG, HT-29, SKOV3, Raji, SGC-7901; Cellular Location: Endoplasmic reticulum lumen.

This endoplasmic reticulum co-chaperone may play a role in protein folding and translocation across the endoplasmic reticulum membrane. DNAJC10 may act as a co-chaperone for HSPA5.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC10
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Dnajc10
Gene Accession No. Q8IXB1, Q9DC23
Gene Alias 1200006L06Rik; D2Ertd706e; DnaJ (Hsp40) homolog, subfamily C, member 10; DnaJ heat shock protein family (Hsp40) member C10; dnaJ homolog subfamily C member 10; DNAJC10; DnaJ-like protein subfamily C member 10-like protein; Endoplasmic reticulum DNA J domain-containing protein 5; endoplasmic reticulum DnaJ-PDI fusion protein 1; ERdj5; ER-resident protein ERdj5; J domain-containing PDI-like protein; j domain-containing protein disulfide isomerase-like protein; J-domain-containing protein disulfide isomerase-like protein; Jpdi; Macrothioredoxin; MTHr; PDIA19; protein disulfide isomerase family A, member 19; UNQ495/PRO1012; zgc:162218
Gene Symbols Dnajc10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 614-793 of human DNAJC10 (NP_0618541).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54431, 66861
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.