Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100479
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100479 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100479 Supplier Invitrogen™ Supplier No. PA5100479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DNAJC11.

Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC11
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dnajc11
Gene Accession No. Q5U458, Q9NVH1
Gene Alias dJ126A5.1; DnaJ (Hsp40) homolog, subfamily C, member 11; DnaJ heat shock protein family (Hsp40) member C11; dnaJ homolog subfamily C member 11; DNAJC11; E030019A03Rik; novel DnaJ domain-containing protein
Gene Symbols Dnajc11
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAJC11(Accession Q9NVH1), corresponding to amino acid residues K484-V534.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230935, 362666, 55735
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.