Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAJC11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585470
Description
Keep as concentrated solution. Positive Control: 293T. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.
Specifications
DNAJC11 | |
Polyclonal | |
Unconjugated | |
Dnajc11 | |
dJ126A5.1; DnaJ (Hsp40) homolog, subfamily C, member 11; DnaJ heat shock protein family (Hsp40) member C11; dnaJ homolog subfamily C member 11; DNAJC11; E030019A03Rik; novel DnaJ domain-containing protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55735 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.87 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NVH1 | |
Dnajc11 | |
Recombinant protein encompassing a sequence within the center region of human DNAJC11. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction