Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570328
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570328 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570328 Supplier Invitrogen™ Supplier No. PA570328
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 86%; Goat: 82%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 86%; Zebrafish: 85%.

This gene encodes a member of a subclass of the HSP40/DnaJ protein family. Members of this family of proteins are associated with complex assembly, protein folding, and export. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC12
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAJC12
Gene Accession No. Q9R022
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 12; DnaJ heat shock protein family (Hsp40) member C12; dnaJ homolog subfamily C member 12; DnaJ subfamily C member 12-like protein; DNAJC12; DnaJ-like protein subfamily C member 12-like protein; J domain containing protein 1; J domain containing protein 1 (JDP1); J domain protein 1; j domain-containing protein 1; JDP1; mJDP1
Gene Symbols DNAJC12
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of mouse DNAJC12.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 30045
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.