Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC14 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553618
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553618 Supplier Invitrogen™ Supplier No. PA553618
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WGWLELPWVK QNINRQGNAP VASGRYCQPE EEVARLLTMA GVPEDELNPF HVLGVEATAS DVELKKAYRQ LAVMVHPDKN HHPRAEEAFK VLRAAWDIVS NAEKRKEYEM KRMAENELSR SVNEFLSKLQ DDLKEAMNTM MCSRCQG Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 95%.

ER-membrane-associated protein; may be involved in ER export regulation of a GPCR.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC14
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC14
Gene Accession No. Q5XIX0, Q6Y2X3, Q921R4
Gene Alias 5730551F12Rik; DNAJ; DnaJ (Hsp40) homolog, subfamily C, member 14; DnaJ heat shock protein family (Hsp40) member C14; dnaJ homolog subfamily C member 14; DnaJ protein homolog 3; Dnajc14; dopamine receptor interacting protein; dopamine receptor-interacting protein of 78 kDa; DRiP78; HDJ3; hDj-3; Human DnaJ protein 3; LIP6; LYST-interacting protein LIP6
Gene Symbols DNAJC14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJC14. Recombinant protein control fragment (Product #RP-90967).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114481, 74330, 85406
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.