Learn More
Invitrogen™ DNAJC15 Polyclonal Antibody

Description
Peptide sequence: FAGRYAFRIW KPLEQVITET AKKISTPSFS SYYKGGFEQK MSRREAGLIL.
Specifications
Specifications
| Antigen | DNAJC15 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | DNAJC15 |
| Gene Accession No. | Q9Y5T4 |
| Gene Alias | 1110003P16Rik; cell growth-inhibiting gene 22 protein; DnaJ (Hsp40) homolog, subfamily C, member 15; DnaJ (Hsp40) homolog, subfamily D, member 1; DNAJ domain-containing; DnaJ heat shock protein family (Hsp40) member C15; dnaJ homolog subfamily C member 15; DNAJC15; Dnajd1; GIG22; HSD18; MCJ; methylation-controlled J protein |
| Gene Symbols | DNAJC15 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.