Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJC15 Rabbit anti-Human, Unconjugated, Clone: CBC-4, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB19981
Description
DNAJC15 Monoclonal antibody specifically detects DNAJC15 in Human samples. It is validated for WB-Ce, IHC-P.Specifications
DNAJC15 | |
Monoclonal | |
Unconjugated | |
DNAJC15 | |
Rabbit | |
100 μg | |
29103 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Western Blot, Immunohistochemistry (Paraffin) | |
CBC-4 | |
DnaJ (Hsp40) homolog, subfamily C, member 15 | |
DNAJD1, HSD18, MCJ | |
A synthetic peptide corresponding to human DNAJC15 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction