Learn More
Invitrogen™ DNAJC17 Polyclonal Antibody

Description
Antibody detects endogenous levels of total DNAJC17.
Specifications
Specifications
| Antigen | DNAJC17 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | DNAJC17 |
| Gene Accession No. | Q91WT4, Q9NVM6 |
| Gene Alias | 1700025B16Rik; C87112; D9Bwg1371e; DnaJ (Hsp40) homolog, subfamily C, member 17; DnaJ heat shock protein family (Hsp40) member C17; dnaJ homolog subfamily C member 17; Dnajc17; RGD1308015 |
| Gene Symbols | DNAJC17 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.