Learn More
Invitrogen™ DNAJC19 Monoclonal Antibody (3H4)

Description
Peptide Sequence: RYVLQAMKHM EPQVKQVFQS LPKSAFSGGY YRGGFEPKMT KREAALILGV SPTANKGKIR DAHRRIMLLN HPDKGGSPYI AAKINEAKDL LEGQAKK.
Specifications
Specifications
| Antigen | DNAJC19 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 3H4 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Dnajc19 |
| Gene Accession No. | Q96DA6 |
| Gene Alias | 1810055D05Rik; AA959924; DnaJ (Hsp40) homolog, subfamily C, member 19; DnaJ heat shock protein family (Hsp40) member C19; dnaJ homolog subfamily C member 19; DnaJ homolog, subfamily C, member 19; DNAJC19; DnaJ-like protein subfamily C member 19; homolog of yeast TIM14; hypothetical protein LOC513918; hypothetical protein MGC73251; mitochondrial import inner membrane translocase subunit TIM14; PAM18; RGD1560220; TIM14; TIMM14; zgc:73251 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.