Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570454 Supplier Invitrogen™ Supplier No. PA570454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 93%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

The protein encoded by this gene is thought to be part of a complex involved in the ATP-dependent transport of transit peptide-containing proteins from the inner cell membrane to the mitochondrial matrix. Defects in this gene are a cause of 3-methylglutaconic aciduria type 5 (MGA5), also known as dilated cardiomyopathy with ataxia (DCMA). Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1, 2, 6, 10, 14 and 19. [provided by RefSeq, Jan 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC19
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dnajc19
Gene Accession No. Q9CQV7
Gene Alias 1810055D05Rik; AA959924; DnaJ (Hsp40) homolog, subfamily C, member 19; DnaJ heat shock protein family (Hsp40) member C19; dnaJ homolog subfamily C member 19; DnaJ homolog, subfamily C, member 19; DNAJC19; DnaJ-like protein subfamily C member 19; homolog of yeast TIM14; hypothetical protein LOC513918; hypothetical protein MGC73251; mitochondrial import inner membrane translocase subunit TIM14; PAM18; RGD1560220; TIM14; TIMM14; zgc:73251
Gene Symbols Dnajc19
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human DNAJC19 (aa 35-84).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67713
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.