Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5106405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5106405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5106405 Supplier Invitrogen™ Supplier No. PA5106405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total DNAJC19.

The protein encoded by this gene is thought to be part of a complex involved in the ATP-dependent transport of transit peptide-containing proteins from the inner cell membrane to the mitochondrial matrix. Defects in this gene are a cause of 3-methylglutaconic aciduria type 5 (MGA5), also known as dilated cardiomyopathy with ataxia (DCMA). Alternative splicing of this gene results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 1, 2, 6, 10, 14 and 19. [provided by RefSeq, Jan 2012].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC19
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Dnajc19
Gene Accession No. Q96DA6, Q9CQV7
Gene Alias 1810055D05Rik; AA959924; DnaJ (Hsp40) homolog, subfamily C, member 19; DnaJ heat shock protein family (Hsp40) member C19; dnaJ homolog subfamily C member 19; DnaJ homolog, subfamily C, member 19; DNAJC19; DnaJ-like protein subfamily C member 19; homolog of yeast TIM14; hypothetical protein LOC513918; hypothetical protein MGC73251; mitochondrial import inner membrane translocase subunit TIM14; PAM18; RGD1560220; TIM14; TIMM14; zgc:73251
Gene Symbols Dnajc19
Host Species Rabbit
Immunogen A synthesized peptide derived from human DNAJC19(Accession Q96DA6), corresponding to amino acid residues L65-K115.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131118, 502525, 67713
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.