Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109645 Supplier Invitrogen™ Supplier No. PA5109645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MLLLPSAADG RGTAITHALT SASTLCQVEP VGRWFEAFVK RRNRNASASF QELEDKKELS EESEDEELQL EEFPMLKTLD PKDWKNQDHY AVLGLGHVRY KATQRQIKAA HKAMVLKHHP DKRKAAGEPI KEGDNDYFTC.

EIF4EBP1 is one member of a family of translation repressor proteins. It directly interacts with eukaryotic translation initiation factor 4E (eIF4E), which is a limiting component of the multisubunit complex that recruits 40S ribosomal subunits to the 5' end of mRNAs. Interaction of this protein with eIF4E inhibits complex assembly and represses translation. This protein is phosphorylated in response to various signals including UV irradiation and insulin signaling, resulting in its dissociation from eIF4E and activation of mRNA translation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.27 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Dnajc2
Gene Accession No. P54103, Q99543
Gene Alias (rat homolog) mouse id-associated protein 1; AU020218; DnaJ (Hsp40) homolog, subfamily C, member 2; DnaJ heat shock protein family (Hsp40) member C2; dnaJ homolog subfamily C member 2; DnaJ homolog subfamily C member 2, N-terminally processed; Dnajc2; Gliosarcoma-related antigen MIDA1; Id associate 1; Mida1; Mouse Id associate 1; M-phase phosphoprotein 11; MPHOSPH11; MPP11; Unknown (protein for MGC:139161); ZRF1; Zrf2; ZUO1; zuotin related factor 1; zuotin related factor 2; zuotin-related factor 1
Gene Symbols Dnajc2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-140 of human DNAJC2 (NP_055192.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22791, 27000
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.