Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC21 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565550
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565550 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565550 Supplier Invitrogen™ Supplier No. PA565550
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KNHEKSKKHRE MVALLKQQLE EEEENFSRPQ IDENPLDDNS EEEMEDAPKQ KLSKKQKKKK QKPAQNYDDN FNVNGPGEGV KVDPEDTN Highest antigen sequence identity to the following orthologs - mouse 80%, rat 74%.

May act as a co-chaperone for HSP70. May play a role in ribosomal RNA (rRNA) biogenesis, possibly in the maturation of the 60S subunit. Binds the precursor 45S rRNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC21
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC21
Gene Accession No. Q5F1R6
Gene Alias 4930461P20Rik; 9930116P15Rik; DnaJ (Hsp40) homolog, subfamily C, member 21; DnaJ heat shock protein family (Hsp40) member C21; dnaJ homolog subfamily A member 5; DnaJ homolog subfamily C member 21; DnaJ homology subfamily A member 5; dnaja5; dnajc21; fa66c02; GS3; JJJ1; JJJ1 DnaJ domain protein homolog; Protein GS3; putative regulation protein GS3; Unknown (protein for MGC:63563); wu:fa66c02; zgc:63563
Gene Symbols DNAJC21
Host Species Rabbit
Immunogen Recombinant Human DNAJC21. Recombinant protein control fragment (Product #RP-106222).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134218
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.