Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC22 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569646 Supplier Invitrogen™ Supplier No. PA569646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 86%; Dog: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 86%; Rat: 93%.

May function as a co-chaperone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC22
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAJC22
Gene Accession No. Q8CHS2
Gene Alias 2810451A06Rik; AI506245; DnaJ (Hsp40) homolog, subfamily B, member 3; heat shock protein, DNAJ-like 3; DnaJ (Hsp40) homolog, subfamily C, member 22; DnaJ heat shock protein family (Hsp40) member C22; dnaJ homolog subfamily C member 22; Dnajc22; RGD1311098; wurst homolog; wus
Gene Symbols DNAJC22
Host Species Rabbit
Immunogen synthetic peptide corresponding to a region of mouse DNAJC22 (aa 120-169).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72778
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.