Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555232
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555232 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555232 Supplier Invitrogen™ Supplier No. PA555232
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LQRCEDDLRN VGPVDAQVYL EEMSWNEGDH SFYLSCRCGG KYSVSKDEAE EVSLISCDTC SLIIELLHY Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 78%.

Stimulates the ATPase activity of several Hsp70-type chaperones. This ability is enhanced by iron-binding. The iron- bound form is redox-active and can function as electron carrier. Plays a role in the diphthamide biosynthesis, a post-translational modification of histidine which occurs in translation elongation factor 2 (EEF2) which can be ADP-ribosylated by diphtheria toxin and by Pseudomonas exotoxin A (Eta).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC24
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC24
Gene Accession No. Q6P3W2
Gene Alias 1700030A21Rik; 2610027M02Rik; AV066965; AW240712; CSL-type zinc finger-containing protein 3; diphthamide biosynthesis protein 4; DnaJ (Hsp40) homolog, subfamily C, member 24; DnaJ heat shock protein family (Hsp40) member C24; dnaJ homolog subfamily C member 24; Dnajc24; DPH4; DPH4 homolog (JJJ3, S. cerevisiae); DPH4, JJJ3 homolog; j domain protein DjC7; JJJ3; MmDjC7; ZCSL3; zinc finger, CSL domain containing 3; zinc finger, CSL-type containing 3
Gene Symbols DNAJC24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJC24. Recombinant protein control fragment (Product #RP-92431).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120526
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.