Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109646
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109646 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109646 Supplier Invitrogen™ Supplier No. PA5109646
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAVEQMPKKD WYSILGADPS ANISDLKQKY QKLILMYHPD KQSTDVPAGT VEECVQKFIE IDQAWKILGN EETKREYDLQ RCEDDLRNVG PVDAQVYLEE MSWNEGDHSF YLSCRCGGKY SVSKDEAEEV SLISCDTCSL IIELLHYN.

Stimulates the ATPase activity of several Hsp70-type chaperones. This ability is enhanced by iron-binding. The iron- bound form is redox-active and can function as electron carrier. Plays a role in the diphthamide biosynthesis, a post-translational modification of histidine which occurs in translation elongation factor 2 (EEF2) which can be ADP-ribosylated by diphtheria toxin and by Pseudomonas exotoxin A (Eta).
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC24
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene DNAJC24
Gene Accession No. Q91ZF0
Gene Alias 1700030A21Rik; 2610027M02Rik; AV066965; AW240712; CSL-type zinc finger-containing protein 3; diphthamide biosynthesis protein 4; DnaJ (Hsp40) homolog, subfamily C, member 24; DnaJ heat shock protein family (Hsp40) member C24; dnaJ homolog subfamily C member 24; Dnajc24; DPH4; DPH4 homolog (JJJ3, S. cerevisiae); DPH4, JJJ3 homolog; j domain protein DjC7; JJJ3; MmDjC7; ZCSL3; zinc finger, CSL domain containing 3; zinc finger, CSL-type containing 3
Gene Symbols DNAJC24
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 2-149 of human DNAJC24 (NP_859057.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 99349
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.