Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553929
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553929 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553929 Supplier Invitrogen™ Supplier No. PA553929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NIKGKEYGEE ERLYIIRKSM KMSKSQFDSL EDHQKETFLK RELWIKENYE VYKQEQEEEL KKKLANDPRW KRYRRWMKNE GPGRLTFVDD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

This gene represents naturally-occurring mRNAs that are co-transcribed products of the neighboring DNAJC25 and GNG10 genes. These transcripts include the first exon of DNAJC25 and the last two exons of GNG10, resulting in a protein that combines the N-terminus of DNAJC25 and the C-terminus of GNG10. [provided by RefSeq, Dec 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC25
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DNAJC25
Gene Accession No. Q9H1X3
Gene Alias 2010109C08Rik; 2010203O07Rik; bA16L21.2.1; DnaJ (Hsp40) homolog, subfamily C , member 25; DnaJ (Hsp40) homolog, subfamily C, member 25; DnaJ heat shock protein family (Hsp40) member C25; dnaJ homolog subfamily C member 25; DNAJC25; DnaJ-like protein; dnaJ-like protein subfamily c member 25-like protein; ERJ7; GNG10; si:ch211-154e10.2
Gene Symbols DNAJC25
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJC25. Recombinant protein control fragment (Product #RP-102372).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 548645
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.