Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC25 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569340 Supplier Invitrogen™ Supplier No. PA569340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 93%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 77%.

This gene represents naturally-occurring mRNAs that are co-transcribed products of the neighboring DNAJC25 and GNG10 genes. These transcripts include the first exon of DNAJC25 and the last two exons of GNG10, resulting in a protein that combines the N-terminus of DNAJC25 and the C-terminus of GNG10. [provided by RefSeq, Dec 2008].
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC25
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene DNAJC25
Gene Accession No. A2ALW5
Gene Alias 2010109C08Rik; 2010203O07Rik; bA16L21.2.1; DnaJ (Hsp40) homolog, subfamily C , member 25; DnaJ (Hsp40) homolog, subfamily C, member 25; DnaJ heat shock protein family (Hsp40) member C25; dnaJ homolog subfamily C member 25; DNAJC25; DnaJ-like protein; dnaJ-like protein subfamily c member 25-like protein; ERJ7; GNG10; si:ch211-154e10.2
Gene Symbols DNAJC25
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human DNAJC25 (aa 173-222).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72429
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.