Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DNAJC25 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989787
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9787 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9787 Supplier Bioss Supplier No. BS14389R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene represents naturally-occurring mRNAs that are co-transcribed products of the neighboring DNAJC25 and GNG10 genes. These transcripts include the first exon of DNAJC25 and the last two exons of GNG10, resulting in a protein that combines the N-terminus of DNAJC25 and the C-terminus of GNG10. [provided by RefSeq, Dec 2008].

Specifications

Antigen DNAJC25
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene DNAJC25
Gene Accession No. Q9H1X3
Gene Alias 2010109C08Rik; 2010203O07Rik; bA16L21.2.1; BOS_9337; DnaJ (Hsp40) homolog, subfamily C , member 25; DnaJ (Hsp40) homolog, subfamily C, member 25; DnaJ heat shock protein family (Hsp40) member C25; DnaJ homolog subfamily C member 25; DNAJC25; DnaJ-like protein; ERJ7; GNG10; si:ch211-154e10.2
Gene Symbols DNAJC25
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human DNAJC25.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 548645
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.