Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC27 Monoclonal Antibody (3C1)
GREENER_CHOICE

Catalog No. PIMA521894
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521894 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521894 Supplier Invitrogen™ Supplier No. MA521894
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MEANMPKRKE PGRSLRIKVI SMGNAEVGKS CIIKRYCEKR FVSKYLATIG IDYGVTKVHV RDREIKVNIF DMAGHPFFYE VRNEFYKDTQ GVILVYDVGQ KDSFDALDAW LAEMKQELGP HGNMENIIFV VCANKIDCTK HRCVDESEGR LWAESKGFLY FETSAQTGEG INEMFQTFYI SIVDLCENGG KRPTTNSSAS FTKEQADAIR RIRNSKDSWD MLGVKPGASR DEVNKAYRKL AVLLHPDKCV APGSEDAFKA VVNARTALLK NIK.

GTPase which can activate the MEK/ERK pathway and induce cell transformation when overexpressed. May act as a nuclear scaffold for MAPK1, probably by association with MAPK1 nuclear export signal leading to enhanced ERK1/ERK2 signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC27
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3C1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Dnajc27
Gene Accession No. Q9NZQ0
Gene Alias AI639580; APAH1; C330021A05Rik; DnaJ (Hsp40) homolog, subfamily C, member 27; DnaJ heat shock protein family (Hsp40) member C27; dnaJ homolog subfamily C member 27; dnajc27; LOW QUALITY PROTEIN: dnaJ homolog subfamily C member 27; rab and DnaJ domain containing; rab and DnaJ domain-containing protein; Rabj; RabJS; Rab-related GTP-binding protein; Ras-associated protein Rap1; RBJ; zgc:194263; zgc:194279
Gene Symbols Dnajc27
Host Species Mouse
Immunogen DNAJC27 (AAH34049, 1 a.a. ~ 273 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51277
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.