Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557830
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557830 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557830 Supplier Invitrogen™ Supplier No. PA557830
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GGKRPTTNSS ASFTKEQADA IRRIRNSKDS WDMLGVKPGA SRDEVNKAYR KLAVLLHPDK CVAPGSEDAF KAVVNARTAL LK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 91%.

GTPase which can activate the MEK/ERK pathway and induce cell transformation when overexpressed. May act as a nuclear scaffold for MAPK1, probably by association with MAPK1 nuclear export signal leading to enhanced ERK1/ERK2 signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC27
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Dnajc27
Gene Accession No. Q9NZQ0
Gene Alias AI639580; APAH1; C330021A05Rik; DnaJ (Hsp40) homolog, subfamily C, member 27; DnaJ heat shock protein family (Hsp40) member C27; dnaJ homolog subfamily C member 27; dnajc27; LOW QUALITY PROTEIN: dnaJ homolog subfamily C member 27; rab and DnaJ domain containing; rab and DnaJ domain-containing protein; Rabj; RabJS; Rab-related GTP-binding protein; Ras-associated protein Rap1; RBJ; zgc:194263; zgc:194279
Gene Symbols Dnajc27
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DNAJC27. Recombinant protein control fragment (Product #RP-96003).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51277
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.