Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC27 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA570259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA570259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA570259 Supplier Invitrogen™ Supplier No. PA570259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

GTPase which can activate the MEK/ERK pathway and induce cell transformation when overexpressed. May act as a nuclear scaffold for MAPK1, probably by association with MAPK1 nuclear export signal leading to enhanced ERK1/ERK2 signaling.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC27
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Dnajc27
Gene Accession No. Q8CFP6
Gene Alias AI639580; APAH1; C330021A05Rik; DnaJ (Hsp40) homolog, subfamily C, member 27; DnaJ heat shock protein family (Hsp40) member C27; dnaJ homolog subfamily C member 27; dnajc27; LOW QUALITY PROTEIN: dnaJ homolog subfamily C member 27; rab and DnaJ domain containing; rab and DnaJ domain-containing protein; Rabj; RabJS; Rab-related GTP-binding protein; Ras-associated protein Rap1; RBJ; zgc:194263; zgc:194279
Gene Symbols Dnajc27
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human DNAJC27 (aa 52-101).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217378
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.