Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ DNAJC3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583458
Description
Immunogen sequence: YKISTLYYQL GDHELSLSEV RECLKLDQDH KRCFAHYKQV KKLNKLIESA EELIRDGRYT DATSKYESVM KTEPSIAEYT VR.
This gene encodes a protein with multiple tetratricopeptide repeat (TPR) motifs as well as the highly conserved J domain found in DNAJ chaperone family members. It is a member of the tetratricopeptide repeat family of proteins and acts as an inhibitor of the interferon-induced, dsRNA-activated protein kinase (PKR).
Specifications
| DNAJC3 | |
| Polyclonal | |
| Unconjugated | |
| DNAJC3 | |
| AA408985; ACPHD; AU067833; DnaJ (Hsp40) homolog, subfamily C, member 3; DnaJ (Hsp40) homolog, subfamily C, member 3A; DnaJ (Hsp40) homolog, subfamily C, member 3B; DnaJ heat shock protein family (Hsp40) member C3; dnaJ homolog subfamily C member 3; DNAJC3; Dnajc3a; Dnajc3b; Endoplasmic reticulum DNA J domain-containing protein 6; ERdj6; ER-resident protein ERdj6; FLJ21288; HP58; Interferon-induced, double-stranded RNA-activated protein kinase inhibitor; mp58; p58; p58IPK; PRKRI; Protein kinase inhibitor of 58 kDa; protein kinase inhibitor p58; protein kinase, interferon inducible double stranded RNA dependent inhibitor; protein-kinase, interferon-inducible double stranded RNA dependent inhibitor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5611 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q13217 | |
| DNAJC3 | |
| Recombinant protein corresponding to Human DNAJC3. Recombinant protein control fragment (Product #RP-97911). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction