Learn More
DNAJC3 Rabbit anti-Human, Mouse, Polyclonal, ProteinTech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 267211AP150UL

Description
Aliquoting is unnecessary for -20°C storage.
DNAJC3 Polyclonal antibody specifically detects DNAJC3 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
DNAJC3 | |
Polyclonal | |
Unconjugated | |
DNAJC3 | |
AA408985; ACPHD; AU067833; DnaJ (Hsp40) homolog, subfamily C, member 3; DnaJ (Hsp40) homolog, subfamily C, member 3A; DnaJ (Hsp40) homolog, subfamily C, member 3B; DnaJ heat shock protein family (Hsp40) member C3; dnaJ homolog subfamily C member 3; DNAJC3; Dnajc3a; Dnajc3b; Endoplasmic reticulum DNA J domain-containing protein 6; ERdj6; ER-resident protein ERdj6; FLJ21288; HP58; Interferon-induced, double-stranded RNA-activated protein kinase inhibitor; mp58; p58; p58IPK; PRKRI; Protein kinase inhibitor of 58 kDa; protein kinase inhibitor p58; protein kinase, interferon inducible double stranded RNA dependent inhibitor; protein-kinase, interferon-inducible double stranded RNA dependent inhibitor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100037258, 5611 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
760 μg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q13217, Q91YW3 | |
DNAJC3 | |
DNAJC3 Fusion Protein Ag24966 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.