Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DNAJC30 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300191R
Description
DNAJC30 Polyclonal specifically detects DNAJC30 in Human samples. It is validated for ELISA.Specifications
| DNAJC30 | |
| Polyclonal | |
| ELISA | |
| DnaJ (Hsp40) homolog, subfamily C, member 30, dnaJ homolog subfamily C member 30, MGC12943, WBSCR18, Williams Beuren syndrome chromosome region 18, Williams-Beuren syndrome chromosomal region 18 protein | |
| Produced in rabbits immunized with purified, recombinant DNAJC30 (Q96LL9; Ser39-Gly124). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84277 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction