Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC30 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598729
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598729 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598729 Supplier Invitrogen™ Supplier No. PA598729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

DNAJC30 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This intronless gene encodes a member of the DNAJ molecular chaperone homology domain-containing protein family. This gene is deleted in Williams syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at 7q11.23.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC30
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene DNAJC30
Gene Accession No. Q96LL9
Gene Alias DnaJ (Hsp40) homolog, subfamily C, member 30; DnaJ heat shock protein family (Hsp40) member C30; dnaJ homolog subfamily C member 30; DnaJ homolog subfamily C member 30, mitochondrial; DNAJC30; WBSCR18; Williams Beuren syndrome chromosome region 18; williams-Beuren syndrome chromosomal region 18 protein
Gene Symbols DNAJC30
Host Species Rabbit
Immunogen Recombinant Human DnaJ homolog subfamily C member 30 protein (1-226AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84277
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.