Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DNAJC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119173
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119173 200 μL
PIPA5122031 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5119173 Supplier Invitrogen™ Supplier No. PA5119173
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

DNAJC4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen DNAJC4
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene DNAJC4
Gene Accession No. Q9NNZ3
Gene Alias 2010301J22Rik; DANJC4; DnaJ (Hsp40) homolog, subfamily C, member 4; DnaJ heat shock protein family (Hsp40) member C4; dnaJ homolog subfamily C member 4; DNAJC4; DnaJ-like protein HSPF2; heat shock 40kD protein 2; Hspf2; MCG18; multiple endocrine neoplasia type 1 candidate protein 18; multiple endocrine neoplasia type 1 candidate protein number 18; Multiple endocrine neoplasia type 1 candidate protein number 18 homolog
Gene Symbols DNAJC4
Host Species Rabbit
Immunogen Recombinant protein Heat Shock 40kDa Protein 2. The antigen corresponds to amino acid range 1-150 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100522005, 3338
Target Species Human, Pig
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.